Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human VEGFR1/Flt-1 Quantikine ELISA Kit
GSA_VASDP

Catalog No. SVR100C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Pk. (6 Plates)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SVR100C 1 Pk. (6 Plates)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. SVR100C Supplier R&D Systems™ Supplier No. SVR100C
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human VEGFR1/Flt-1 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human VEGFR1/Flt-1 Quantikine ELISA Kit quantifies human VEGFR1/Flt-1 in cell culture supernates (100 ul), cell lysates (10 ul), serum (100 ul), edta plasma (100 ul), heparin plasma (100 ul), saliva (100 ul).

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Assay Range 31.30 to 2000 pg/mL
Assay Sensitivity 8.46 pg/mL
Sample Type Cell Culture Supernatant, Cell Lysates, Plasma (EDTA, Heparin), Saliva, Serum
Specificity Natural and Recombinant Human soluble VEGF R1
For Use With (Application) ELISA
Gene ID (Entrez) 2321
Host Species Human
Quantity 1 Pk. (6 Plates)
Sample Volume Cell Culture Supernates (100 uL),Cell Lysates (10 uL),Serum (100 uL),EDTA Plasma (100 uL),Heparin Plasma (100 uL),Saliva (100 uL)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym EC 2.7.10, EC 2.7.10.1, FLT, FLT1, FLT-1, Fms-like tyrosine kinase 1, fms-related tyrosine kinase 1 (vascular endothelial growth factor/vascularpermeability factor receptor), FRT, Tyrosine-protein kinase FRT, Tyrosine-protein kinase receptor FLT, vascular endothelial growth factor receptor 1, Vascular permeability factor receptor, VEGF R1, VEGFR1, VEGFR-1
Target VEGFR1/Flt-1
Total Assay Time 4.5 hr.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.