Learn More
ACROBiosystems Human VEGF R2 25tests
Supplier:  ACROBiosystems VE2HA22425TESTS
Encompass_Preferred
APC-Labeled Human VEGF R2 Protein His Tag (VE2-HA224) is produced via conjugation of APC to Human VEGF R2 Protein His Tag with a new generation site-specific technology under optimal conditions with a proprietary technology. Human VEGF R2 Protein His Tag is expressed from human 293 cells (HEK293). It contains AA Ala 20 - Glu 764 (Accession # AAI31823.1).
Catalog No. 50-253-4003
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.