Learn More
Description
Specifications
Specifications
| Assay Range | 78 - 5000pg/mL |
| Assay Sensitivity | 7.0pg/mL |
| Product Type | ELISA Kit |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum |
| Specificity | The assay detects both natural and recombinant human VEGFR2/KDR/Flk-1. Cross reactivity and interference of circulating factors of the immune system was evaluated by spiking these proteins at physiologically relevant concentrations into a human VEGFR2/KDR/Flk-1 positive sample. There was no cross reactivity detected, notably not with VEGF-R1 at physiological concentrations and not with VEGF-R3/FLT-4. There was no interference detected, notably not for VEGF-A and VEGF-C. |
| Target Species | Human |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 3791 |
| Gene Symbol | KDR |
| Kit Contents | Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to human VEGF-R2/KDR, Biotin-Conjugate anti-human VEGF-R2/KDR monoclonal antibody, Streptavidin-HRP, Human VEGF-R2/KDR Standard lyophilized, 10ng/mL upon reconstitution, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
