Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human VEGFR2/KDR Quantikine ELISA Kit
SDP

Catalog No. SVR200 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Plate
6 Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SVR200 6 Plates
DVR200 1 Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. SVR200 Supplier R&D Systems™ Supplier No. SVR200

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Human VEGFR2/KDR Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human VEGFR2/KDR Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human VEGFR2/KDR/Flk-1 in cell culture supernates (100 ul), cell lysates (100 ul), serum (20 ul), edta plasma (20 ul), heparin plasma (20 ul).

Specifications

Assay Range 78.10 - 5000pg/mL
Assay Sensitivity 11.4pg/mL
Conjugate HRP
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Line R&D Systems
Sample Type Cell Culture Supernatant, Cell Lysates, Plasma (EDTA, Heparin), Serum
Specificity Natural and Recombinant Human VEGF R2
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 3791
Quantity 6 Plates
Sample Volume Cell Culture Supernates (100uL),Cell Lysates (100uL),Serum (20uL),EDTA Plasma (20uL),Heparin Plasma (20uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym CD309, CD309 antigen, EC 2.7.10, EC 2.7.10.1, Fetal liver kinase 1, fetal liver kinase-1, Flk1, Flk-1, FLK1tyrosine kinase growth factor receptor, KDR, Kinase insert domain receptor, kinase insert domain receptor (a type III receptor tyrosine kinase), KRD1, Ly73, Protein-tyrosine kinase receptor flk-1, soluble VEGFR2, vascular endothelial growth factor receptor 2, VEGF R2, VEGFR, VEGFR2, VEGFR-2
Target VEGFR2/KDR/Flk-1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.