Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human VEGFR3/Flt-4 ELISA Kit (Colorimetric)
SDP

Catalog No. NBP184822 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP184822 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP184822 Supplier Novus Biologicals™ Supplier No. NBP184822
Add to Cart
Add to Cart

ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.

The Human VEGFR3/Flt-4 ELISA Kit (Colorimetric) measures human VEGFR3/Flt-4 in cell culture supernatant, serum, plasma (citrate).

Specifications

Assay Range 0.6 - 40.0ng/mL
Assay Sensitivity 0.03ng/mL
Product Type ELISA Kit
Sample Type Cell Culture Supernatant, Plasma (Citrate), Serum
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 2324
Gene Symbol FLT4
Kit Contents Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to human VEGF-R3/FLT-4, Biotin-Conjugate anti-human VEGF-R3/FLT-4 monoclonal antibody, Streptavidin-HRP, Human VEGF-R3/FLT-4 Standard lyophilized, 80ng/mL upon reconstitution, Sample Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
Quantity 96 Tests
Research Discipline Angiogenesis, Cancer, Hypoxia, Protein Kinase
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym EC 2.7.10, EC 2.7.10.1, FLT-4, fms-related tyrosine kinase 4, LMPH1A, PCLFLT41, soluble VEGFR3 variant 1, soluble VEGFR3 variant 2, soluble VEGFR3 variant 3, Tyrosine-protein kinase receptor FLT4, vascular endothelial growth factor receptor 3, VEGFR-3, VE
Target VEGF R3/Flt-4
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.