Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human VEGF (Research Grade Bevacizumab Biosimilar) Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7158808
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB9947SP 25 μg
MAB9947100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB9947SP Supplier R&D Systems Supplier No. MAB9947SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human Monoclonal Antibody

VEGF Monoclonal specifically detects VEGF in Human samples. It is validated for Neutralization.

Specifications

Antigen VEGF
Applications Neutralization
Classification Monoclonal
Clone Hu101
Conjugate Unconjugated
Dilution Neutralization 10-80 ng/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Alias MVCD1, VAS, vascular endothelial growth factor A, Vascular permeability factor, Vasculotropin, VEGFA, VEGF-A, VEGFMGC70609, VPF, VPFvascular endothelial growth factor
Host Species Human
Immunogen Human VEGF-A
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 7422.0
Test Specificity Detects human VEGF based on Bevacizumab therapeutic antibody. This non-therapeutic antibody uses the same variable region sequence as the therapeutic antibody Bevacizumab. This product is for research use only.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C, as supplied. 1 month, 2 to 8 °C under sterile conditions after opening. 6 months, -20 to -70 °C under sterile conditions after opening.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.