Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human VEGFA (165 a.a.) Full-length Recombinant Protein with His tag expressed in Barley grain

Catalog No. 89966998 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-966-998 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-966-998 Supplier Abnova™ Supplier No. P5816
Add to Cart
Add to Cart

Used for Func, SDS-PAGE, WB-Re

This gene is a member of the PDGF/VEGF growth factor family and encodes a protein that is often found as a disulfide linked homodimer. This protein is a glycosylated mitogen that specifically acts on endothelial cells and has various effects, including mediating increased vascular permeability, inducing angiogenesis, vasculogenesis and endothelial cell growth, promoting cell migration, and inhibiting apoptosis. Elevated levels of this protein is linked to POEMS syndrome, also known as Crow-Fukase syndrome. Mutations in this gene have been associated with proliferative and nonproliferative diabetic retinopathy. Alternatively spliced transcript variants, encoding either freely secreted or cell-associated isoforms, have been characterized. There is also evidence for the use of non-AUG (CUG) translation initiation sites upstream of, and in-frame with the first AUG, leading to additional isoforms. [provided by RefSeq]

Specifications

Accession Number NP_001165097
Concentration 100μg/mL
For Use With (Application) Functional Assay, SDS-PAGE, Western Blot
Formulation Lyophilized from PBS, pH 7.2
Gene ID (Entrez) 7422
Molecular Weight (g/mol) 30kDa
Name Human VEGFA (165 a.a.) Full-length Recombinant Protein with His tag expressed in Barley grain
Purification Method Chromatography
Quality Control Testing SDS-PAGE Stained with Coomassie Blue
Quantity 1 mg
Storage Requirements Store at −20°C on dry atmosphere.After reconstitution with sterile water to a concentration of no less than 100 ug/mL, store at −20°C. For long term storage it is recommended to add a carrier protein (0.1% HSA or BSA).Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Endotoxin Concentration <0.005ng/μg protein (0.05EU/μg) as measured by turbidimetric kinetic assay
Gene Alias MGC70609, VEGF, VEGF-A, VPF
Common Name VEGFA
Gene Symbol VEGFA
Biological Activity The ED50 is determined by Cell Proliferation Assay for the dose-dependent effects of serial dilutions of recombinant human VEGFA on HUVEC cells. The ED50 value was determined to be 0.58ng/mL.
Species Plants
Recombinant Recombinant
Protein Tag His Tag
Expression System Barley grain (Hordeum vulgare) expression system
Form Lyophilized
Purity or Quality Grade >95% by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.