Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human VEGFB (P49765, 167 amino acids) Partial Recombinant Protein

Catalog No. 89967010 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-967-010 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-967-010 Supplier Abnova™ Supplier No. P4113

Used for Func, SDS-PAGE

Specifications

Accession Number P49765
For Use With (Application) Functional Study, SDS-PAGE
Formulation Lyophilized
Gene ID (Entrez) 7423
Molecular Weight (g/mol) 19.63kDa
Name VEGFB (Human) Recombinant Protein
Preparation Method Escherichia coli expression system
Quantity 20 μg
Storage Requirements Store at -20°C on dry atmosphere. After reconstitution with 0.1% acetic acid, store at -20°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Endotoxin Concentration <0.1ng/μg
Gene Alias VEGFL/VRF
Common Name VEGFB
Gene Symbol VEGFB
Biological Activity Per researcher's feedback, the protein is biologically active as determined by the dose-dependent stimulation of the proliferation of human umbilical vein endothelial cells (HUVEC) in the presence of human VEGF165. The expected ED50 for this effect is 0.5
Species E. coli
Recombinant Recombinant
Protein Tag None
Expression System Escherichia coli expression system
Form Lyophilized
Purity or Quality Grade ≥98% by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.