Learn More
ACROBiosystems Recombinant Protein;Human VEGF121 Protein, His Tag;HEK293;50UG;VE1,RP1-261G23.1,MGC70609,MVCD1,MVCD-1,VEGF 165,VEGF-A,VEGFA,VPF,VEGF165,VEGF-165,VEGF121,VEGF120,VEGF186,VEGF164,VEGF110
Supplier:  ACROBiosystems VE1H524650UG
Encompass_Preferred
Recombinant Protein;Human VEGF121 Protein, His Tag;50UG;Human VEGF121, His Tag (VE1-H5246) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 147 (Accession # P15692-9).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 15.0 kDa. As a result of glycosylation, the protein migrates as 17 kDa and 20 kDa under reducing (R) condition, and 35-45 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Human VEGFR1/R2, Fc Tag at 1 μg/mL (100 μL/well) can bind Human VEGF121, His Tag (Cat. No. VE1-H5246) with a linear range of 2-40 ng/mL (QC tested).
Catalog No. 50-108-9754
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.