Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ProSci Human Vein Lysate
SDP

Catalog No. 16818101
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-818-101 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-818-101 Supplier ProSci Supplier No. XBL110520100
Add to Cart
Add to Cart

Total protein normal vein - Human

  • Human vein tissue lysate was prepared by homogenization using a proprietary technique
  • The tissue was frozen in liquid nitrogen immediately after excision and then stored at -70°C
  • The human vein tissue total protein is provided in a buffer including HEPES (pH7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors
  • For quality control purposes, the vein tissue pattern on SDS-PAGE gel is shown to be consistent for each lot by visualization with coomassie blue staining
  • The vein tissue is then Western analyzed by either GAPDH or β-actin antibody, and the expression level is consistent with each lot.

Specifications

Tissue Vein
Quantity 0.1 mg
Tissue Condition Normal
Host Species Human
Concentration 2 mg/mL
Protein State Denatured
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.