Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human VGF Synthetic Peptide

Catalog No. PEP0643
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP0643 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP0643 Supplier Invitrogen™ Supplier No. PEP0643
Add to Cart
Add to Cart

Synthetic Peptide

VGF Synthetic Peptide, PEP-0643, from Invitrogen.

VGF was initially identified as a rapidly regulated gene product in nerve growth factor-treated PC12 cells. This protein is synthesized in neurons in the central and peripheral nervous system as well as in the pituitary, adrenal medulla, endocrine cells of the stomach, and pancreatic beta cells. VGF is thought to be involved in organism energy balance and regulation of homeostasis as mice lacking this gene show deficits in these areas. More recent results suggest that VGF is upregulated by brain-derived neurotrophic factor (BDNF) and can stimulate the proliferation of hippocampal progenitor cells and produce antidepressant-like behavioral effects, suggesting that this pathway may be a suitable target for therapeutic treatments.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 7425
Species Human
Purification Method purified
Accession Number O15240
Common Name Human VGF
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Symbol VGF
Lyophilized No
Preservative 0.02% sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.