Learn More
Description
- Also known as: RCA1; VHL1; pVHL; HRCA1
- Von Hippel Lindau, VHL, is a component of the protein complex that includes elongin B, elongin C, and cullin-2, and possesses ubiquitin ligase E3 activity.
- VHL is involved in the ubiquitination and degradation of hypoxia-inducible-factor (HIF).
- RNA polymerase II subunit POLR2G/RPB7 is also reported to be a target of VHL.
- VHL is a dominantly inherited familial cancer syndrome predisposing to a variety of malignant and benign tumors.
- Alternatively spliced transcripts encoding different proteins have been described.
- The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | hVHL |
| Accession Number | NP_000542 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 655 bp |
| Gene ID (Entrez) | 7428 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
