Learn More
Description
Specifications
Specifications
| Assay Range | 6.25 - 100ng/mL |
| Assay Sensitivity | 0.338ng/mL |
| Product Type | Human Vitamin D BP Quantikine™ ELISA Kit |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Urine |
| Specificity | Natural and Recombinant Human Vitamin D BP |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 2638 |
| Kit Contents | Natural human Vitamin D Binding Protein Standard |
| Sample Volume | Cell Culture Supernates (50uL),Serum (10uL),EDTA Plasma (10uL),Heparin Plasma (10uL),Urine (50uL) |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
| Show More |
For Research Use Only. Not for use in Humans.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
