Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human VNN3 Partial ORF (NP_060869.2, 175 a.a. - 274 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89967084 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-967-084 25 μg
89-967-083 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-967-084 Supplier Abnova™ Supplier No. H00055350Q01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene is the central gene in a cluster of three vanin genes on chromosome 6q23-q24. The open reading frame is disrupted by a frameshift, and all splice variants that have been described are candidates for nonsense-mediated decay (NMD). Consequently, it is unlikely that this gene expresses a protein in vivo, so it is classified as a pseudogene. Extensive alternative splicing has been described; the two most common variants are represented as RefSeqs. [provided by RefSeq]

Sequence: ARYHKYNLFAPEIQFDFPKDSELVTFDTPFGKFGIFTCFDIFSHDPAVVVVDEFQLTAFSTPQHGTTRCPSSRLFPSIQHGPRPWESIYLLQIPTTPACT

Specifications

Accession Number NP_060869.2
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 55350
Molecular Weight (g/mol) 36.63kDa
Name VNN3 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen ARYHKYNLFAPEIQFDFPKDSELVTFDTPFGKFGIFTCFDIFSHDPAVVVVDEFQLTAFSTPQHGTTRCPSSRLFPSIQHGPRPWESIYLLQIPTTPACT
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias HSA238982/MGC171203/PAGEL-beta/PAGEL-eta/PAGEL-zeta
Common Name VNN3
Gene Symbol VNN3
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.