Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human VPS16 Partial ORF (NP_072097.2, 754 a.a. - 839 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89967097 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-967-097 25 μg
89-967-096 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-967-097 Supplier Abnova™ Supplier No. H00064601Q01L

Used for AP, Array, ELISA, WB-Re

Vesicle mediated protein sorting plays an important role in segregation of intracellular molecules into distinct organelles. Genetic studies in yeast have identified more than 40 vacuolar protein sorting (VPS) genes involved in vesicle transport to vacuoles. This gene encodes the human homolog of yeast class C Vps16 protein. The mammalian class C Vps proteins are predominantly associated with late endosomes/lysosomes, and like their yeast counterparts, may mediate vesicle trafficking steps in the endosome/lysosome pathway. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: IGYLPFVEICMKQHNKYEAKKYASRVGPEQKVKALLLVGDVAQAADVAIEHRNEAELSLVLSHCTGATDGATADKIQRARAQAQKK

Specifications

Accession Number NP_072097.2
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 64601
Molecular Weight (g/mol) 35.2kDa
Name VPS16 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen IGYLPFVEICMKQHNKYEAKKYASRVGPEQKVKALLLVGDVAQAADVAIEHRNEAELSLVLSHCTGATDGATADKIQRARAQAQKK
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias hVPS16
Common Name VPS16
Gene Symbol VPS16
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.