Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human VPS26A Full-length ORF (NP_004887.2, 1 a.a. - 327 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89967110 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-967-110 25 μg
89-967-109 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-967-110 Supplier Abnova™ Supplier No. H00009559P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene belongs to a group of vacuolar protein sorting (VPS) genes. The encoded protein is a component of a large multimeric complex, termed the retromer complex, involved in retrograde transport of proteins from endosomes to the trans-Golgi network. The close structural similarity between the yeast and human proteins that make up this complex suggests a similarity in function. Expression studies in yeast and mammalian cells indicate that this protein interacts directly with VPS35, which serves as the core of the retromer complex. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq]

Sequence: MSFLGGFFGPICEIDIVLNDGETRKMAEMKTEDGKVEKHYLFYDGESVSGKVNLAFKQPGKRLEHQGIRIEFVGQIELFNDKSNTHEFVNLVKELALPGELTQSRSYDFEFMQVEKPYESYIGANVRLRYFLKVTIVRRLTDLVKEYDLIVHQLATYPDVNNSIKMEVGIEDCLHIEFEYNKSKYHLKDVIVGKIYFLLVRIKIQHMELQLIKKEITGIGPSTTTETETIAKYEIMDGAPVKGESIPIRLFLAGYDPTPTMRDVNKKFSVRYFLNLVLVDEEDRRYFKQQEIILWRKAPEKLRKQRTNFHQRFESPESQASAEQPEM

Specifications

Accession Number NP_004887.2
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 9559
Molecular Weight (g/mol) 64.6kDa
Name VPS26A (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MSFLGGFFGPICEIDIVLNDGETRKMAEMKTEDGKVEKHYLFYDGESVSGKVNLAFKQPGKRLEHQGIRIEFVGQIELFNDKSNTHEFVNLVKELALPGELTQSRSYDFEFMQVEKPYESYIGANVRLRYFLKVTIVRRLTDLVKEYDLIVHQLATYPDVNNSIKMEVGIEDCLHIEFEYNKSKYHLKDVIVGKIYFLLVRIKIQHMELQLIKKEITGIGPSTTTETETIAKYEIMDGAPVKGESIPIRLFLAGYDPTPTMRDVNKKFSVRYFLNLVLVDEEDRRYFKQQEIILWRKAPEKLRKQRTNFHQRFESPESQASAEQPEM
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FLJ12930/HB58/Hbeta58/PEP8A/VPS26
Common Name VPS26A
Gene Symbol VPS26A
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.