Learn More
Description
- PIK3C3 phosphatidylinositol 3- kinase catalytic subunit type 3 [ Homo sapiens (human) ]
- Also known as: VPS34; Vps34; hVps34
- The VPS34 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- PIK3C3 is a catalytic subunit of the PI3K complex that mediates formation of phosphatidylinositol 3- phosphate.
- Different complex forms are believed to play a role in multiple membrane trafficking pathways.
- PI3KC3- C1 is involved in initiation of autophagosomes and PI3KC3- C2 in maturation of autophagosomes and endocytosis.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | VPS34 |
| Accession Number | NP_002638 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 2677 bp |
| Gene ID (Entrez) | 5289 |
| Gene Alias | EC 2.7.1, EC 2.7.1.137, hVps34, MGC61518, phosphatidylinositol 3-kinase catalytic subunit type 3, Phosphatidylinositol 3-kinase p100 subunit, Phosphoinositide-3-kinase class 3, phosphoinositide-3-kinase, class 3, PI 3-Kinase C3, PI3K type 3, PI3-kinase type 3, PIK3C3, PtdIns-3-kinase type 3, Vps34 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
