Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human VPS45 Full-length ORF (NP_009190.2, 1 a.a. - 570 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89967144 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-967-144 25 μg
89-967-143 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-967-144 Supplier Abnova™ Supplier No. H00011311P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

Vesicle mediated protein sorting plays an important role in segregation of intracellular molecules into distinct organelles. Genetic studies in yeast have identified more than 40 vacuolar protein sorting (VPS) genes involved in vesicle transport to vacuoles. This gene is a member of the Sec1 domain family, and shows a high degree of sequence similarity to mouse, rat and yeast Vps45. The exact function of this gene is not known, but its high expression in peripheral blood mononuclear cells suggests a role in trafficking proteins, including inflammatory mediators. [provided by RefSeq]

Sequence: MNVVFAVKQYISKMIEDSGPGMKVLLMDKETTGIVSMVYTQSEILQKEVYLFERIDSQNREIMKHLKAICFLRPTKENVDYIIQELRRPKYTIYFIYFSNVISKSDVKSLAEADEQEVVAEVQEFYGDYIAVNPHLFSLNILGCCQGRNWDPAQLSRTTQGLTALLLSLKKCPMIRYQLSSEAAKRLAECVKQVITKEYELFEFRRTEVPPLLLILDRCDDAITPLLNQWTYQAMVHELLGINNNRIDLSRVPGISKDLREVVLSAENDEFYANNMYLNFAEIGSNIKNLMEDFQKKKPKEQQKLESIADMKAFVENYPQFKKMSGTVSKHVTVVGELSRLVSERNLLEVSEVEQELACQNDHSSALQNIKRLLQNPKVTEFDAARLVMLYALHYERHSSNSLPGLMMDLRNKGVSEKYRKLVSAVVEYGGKRVRGSDLFSPKDAVAITKQFLKGLKGVENVYTQHQPFLHETLDHLIKGRLKENLYPYLGPSTLRDRPQDIIVFVIGGATYEEALTVYNLNRTTPGVRIVLGGTTVHNTKSFLEEVLASGLHSRSKESSQVTSRSASRR

Specifications

Accession Number NP_009190.2
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 11311
Molecular Weight (g/mol) 91.5kDa
Name VPS45 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MNVVFAVKQYISKMIEDSGPGMKVLLMDKETTGIVSMVYTQSEILQKEVYLFERIDSQNREIMKHLKAICFLRPTKENVDYIIQELRRPKYTIYFIYFSNVISKSDVKSLAEADEQEVVAEVQEFYGDYIAVNPHLFSLNILGCCQGRNWDPAQLSRTTQGLTALLLSLKKCPMIRYQLSSEAAKRLAECVKQVITKEYELFEFRRTEVPPLLLILDRCDDAITPLLNQWTYQAMVHELLGINNNRIDLSRVPGISKDLREVVLSAENDEFYANNMYLNFAEIGSNIKNLMEDFQKKKPKEQQKLESIADMKAFVENYPQFKKMSGTVSKHVTVVGELSRLVSERNLLEVSEVEQELACQNDHSSALQNIKRLLQNPKVTEFDAARLVMLYALHYERHSSNSLPGLMMDLRNKGVSEKYRKLVSAVVEYGGKRVRGSDLFSPKDAVAITKQFLKGLKGVENVYTQHQPFLHETLDHLIKGRLKENLYPYLGPSTLRDRPQDIIVFVIGGATYEEALTVYNLNRTTPGVRIVLGGTTVHNTKSFLEEVLASGLHSRSKESSQVTSRSASRR
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias H1/H1VPS45/VPS45A/VPS45B/VPS54A/VSP45/VSP45A
Common Name VPS45
Gene Symbol VPS45
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.