Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human VPS4A Full-length ORF (NP_037377.1, 1 a.a. - 437 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89967146 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-967-146 10 μg
89-967-147 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-967-146 Supplier Abnova™ Supplier No. H00027183P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is a member of the AAA protein family (ATPases associated with diverse cellular activities), and is the homolog of the yeast Vps4 protein. In humans, two paralogs of the yeast protein have been identified. The former share a high degree of aa sequence similarity with each other, and also with yeast Vps4 and mouse Skd1 proteins. The mouse Skd1 (suppressor of K+ transport defect 1) has been shown to be really an yeast Vps4 ortholog. Functional studies indicate that both human paralogs associate with the endosomal compartments, and are involved in intracellular protein trafficking, similar to Vps4 protein in yeast. The gene encoding this paralog has been mapped to chromosome 16; the gene for the other resides on chromosome 18. [provided by RefSeq]

Sequence: MTTSTLQKAIDLVTKATEEDKAKNYEEALRLYQHAVEYFLHAIKYEAHSDKAKESIRAKCVQYLDRAEKLKDYLRSKEKHGKKPVKENQSEGKGSDSDSEGDNPEKKKLQEQLMGAVVMEKPNIRWNDVAGLEGAKEALKEAVILPIKFPHLFTGKRTPWRGILLFGPPGTGKSYLAKAVATEANNSTFFSVSSSDLMSKWLGESEKLVKNLFELARQHKPSIIFIDEVDSLCGSRNENESEAARRIKTEFLVQMQGVGNNNDGTLVLGATNIPWVLDSAIRRRFEKRIYIPLPEEAARAQMFRLHLGSTPHNLTDANIHELARKTEGYSGADISIIVRDSLMQPVRKVQSATHFKKVCGPSRTNPSMMIDDLLTPCSPGDPGAMEMTWMDVPGDKLLEPVVCMSDMLRSLATTRPTVNADDLLKVKKFSEDFGQES

Specifications

Accession Number NP_037377.1
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 27183
Molecular Weight (g/mol) 75.3kDa
Name VPS4A (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MTTSTLQKAIDLVTKATEEDKAKNYEEALRLYQHAVEYFLHAIKYEAHSDKAKESIRAKCVQYLDRAEKLKDYLRSKEKHGKKPVKENQSEGKGSDSDSEGDNPEKKKLQEQLMGAVVMEKPNIRWNDVAGLEGAKEALKEAVILPIKFPHLFTGKRTPWRGILLFGPPGTGKSYLAKAVATEANNSTFFSVSSSDLMSKWLGESEKLVKNLFELARQHKPSIIFIDEVDSLCGSRNENESEAARRIKTEFLVQMQGVGNNNDGTLVLGATNIPWVLDSAIRRRFEKRIYIPLPEEAARAQMFRLHLGSTPHNLTDANIHELARKTEGYSGADISIIVRDSLMQPVRKVQSATHFKKVCGPSRTNPSMMIDDLLTPCSPGDPGAMEMTWMDVPGDKLLEPVVCMSDMLRSLATTRPTVNADDLLKVKKFSEDFGQES
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FLJ22197/SKD1/SKD2/VPS4/VPS4-1
Common Name VPS4A
Gene Symbol VPS4A
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.