Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human VPS54 Full-length ORF (AAH41868.1, 1 a.a. - 824 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89967160 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-967-160 25 μg
89-967-159 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-967-160 Supplier Abnova™ Supplier No. H00051542P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes for a protein that in yeast forms part of a trimeric vacuolar-protein-sorting complex that is required for retrograde transport of proteins from prevacuoles to the late Golgi compartment. As in yeast, mammalian Vps54 proteins contain a coiled-coil region and dileucine motifs. Alternative splicing results in multiple transcript variants encoding different isoforms [provided by RefSeq]

Sequence: MLPTKNRIKREKIHERCKNICPPKDTFERTLLHTHDKSRTDLEQVPKIFMKPDFALDDSLTFNSVLPWSHFNTAGGKGNRDAASSKLLQEKLSHYLDIVEVNIAHQISLRSEAFFHAMTSQHELQDYLRKTSQAVKMLRDKIAQIDKVMCEGSLHILRLALTRNNCVKVYNKLKLMATVHQTQPTVQVLLSTSEFVGALDLIATTQEVLQQELQGIHSFRHLGSQLCELEKLIDKMMIAEFSTYSHSDLNRPLEDDCQVLEECVINKVSQTEEIDTDVVVKLADQMRMLNFPQWFDLLKDIFSKFTIFLQRVKATLNIIHSVVLSVLDKNQRTRELEEISQQKNAAKDNSLDTEVAYLIHEGMFISDAFGEGELTPIAVDTTSQRNASPNSEPCSSDSVSEPECTTDSSSSKEHTSSSAIPGGVDIMVSEDMKLTDSELGKLANNIQELLYSASDICHDRAVKFLMSRAKDGFLEKLNSMEFITLSRLMETFILDTEQICGRKSTSLLGALQSQAIKFVNRFHEERKTKLSLLLDNERWKQADVPAEFQDLVDSLSDGKIALPEKKSGATEERKPAEVLIVEGQQYAVVGTVLLLIRIILEYCQCVDNIPSVTTDMLTRLSDLLKYFNSRSCQLVLGAGALQVVGLKTITTKNLALSSRCLQLIVHYIPVIRAHFEARLPPKQYSMLRHFGHITKDYHDHIAEISAKLVAIMDSLFDKLLSKYEVKAPVPSACFRNICKQMTKMHEAIFDHLPEEQTQMLFLRINASYKLHLKKQLSHLNVINDGGPQNGLVTADVAFYTGNLQALKGLKDLDLNMAEIWEQKR

Specifications

Accession Number AAH41868.1
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 51542
Molecular Weight (g/mol) 119.6kDa
Name VPS54 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MLPTKNRIKREKIHERCKNICPPKDTFERTLLHTHDKSRTDLEQVPKIFMKPDFALDDSLTFNSVLPWSHFNTAGGKGNRDAASSKLLQEKLSHYLDIVEVNIAHQISLRSEAFFHAMTSQHELQDYLRKTSQAVKMLRDKIAQIDKVMCEGSLHILRLALTRNNCVKVYNKLKLMATVHQTQPTVQVLLSTSEFVGALDLIATTQEVLQQELQGIHSFRHLGSQLCELEKLIDKMMIAEFSTYSHSDLNRPLEDDCQVLEECVINKVSQTEEIDTDVVVKLADQMRMLNFPQWFDLLKDIFSKFTIFLQRVKATLNIIHSVVLSVLDKNQRTRELEEISQQKNAAKDNSLDTEVAYLIHEGMFISDAFGEGELTPIAVDTTSQRNASPNSEPCSSDSVSEPECTTDSSSSKEHTSSSAIPGGVDIMVSEDMKLTDSELGKLANNIQELLYSASDICHDRAVKFLMSRAKDGFLEKLNSMEFITLSRLMETFILDTEQICGRKSTSLLGALQSQAIKFVNRFHEERKTKLSLLLDNERWKQADVPAEFQDLVDSLSDGKIALPEKKSGATEERKPAEVLIVEGQQYAVVGTVLLLIRIILEYCQCVDNIPSVTTDMLTRLSDLLKYFNSRSCQLVLGAGALQVVGLKTITTKNLALSSRCLQLIVHYIPVIRAHFEARLPPKQYSMLRHFGHITKDYHDHIAEISAKLVAIMDSLFDKLLSKYEVKAPVPSACFRNICKQMTKMHEAIFDHLPEEQTQMLFLRINASYKLHLKKQLSHLNVINDGGPQNGLVTADVAFYTGNLQALKGLKDLDLNMAEIWEQKR
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias HCC8/SLP-8p/VPS54L/hVps54L
Common Name VPS54
Gene Symbol VPS54
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.