Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human VSIG1 DuoSet ELISA
SDP

Catalog No. DY481805 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit (for 5 Plates)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DY481805 1 Kit (for 5 Plates)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DY481805 Supplier R&D Systems™ Supplier No. DY481805
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

R&D Systems™ Human VSIG1 DuoSet ELISA contains the basic components required for the development of sandwich ELISAs to measure natural and recombinant human V-Set and Immunoglobulin domain-containing protein 1 (VSIG1).

The Human VSIG1 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human VSIG1.

Specifications

Assay Range 31.20 to 2000 pg/mL
Product Type Solid Phase Sandwich ELISA
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 340547
Quantity 1 Kit (for 5 Plates)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym Cell surface A33 antigen, dJ889N15.1, Glycoprotein A34, GPA34, MGC44287,1700062D20Rik, V-set and immunoglobulin domain containing 1, V-set and immunoglobulin domain-containing protein 1
Target VSIG1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.