Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human VSIG4 Fc Chimera Recombinant Protein

Catalog No. PIRP88184
Change view
Click to view available options
Quantity:
1 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP88184 1 mg
PIRP88183 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIRP88184 Supplier Invitrogen™ Supplier No. RP88184
Add to Cart
Add to Cart

Recombinant Protein

It is recommended that the protein be aliquoted for optimal storage. It is recommended that sterile water (1588 μL) be added to the vial to prepare a stock solution of 0.25 mg/mL. Concentration is measured by UV.

T cell activation by APCs is positively and negatively regulated by members of the B7 family. VSIG4 is a strong negative regulator of murine and human T cell proliferation and IL-2 production.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9Y279
Concentration 0.63 mg/mL
For Use With (Application) Control, Neutralization
Format Lyophilized
Formulation PBS with 5% mannitol, 5% trehalose, 0.01% Tween 80 and no preservative; pH 7.4
Gene ID (Entrez) 11326
Molecular Weight (g/mol) 57 kDa
Name Human VSIG4 Fc Chimera
pH Range 7.4
Purification Method Purified
Quantity 1 mg
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Endotoxin Concentration <1.0 EU/μg
Gene Alias A530061A11; BC025105; complement receptor of the immunoglobulin superfamily; CRIg; Ig superfamily protein; MGC137243 protein; protein Z39Ig; UNQ317/PRO362; V-set and immunoglobulin domain containing 4; V-set and immunoglobulin domain-containing protein 4; VSIG4; Z39IG
Common Name VSIG4
Gene Symbol VSIG4
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag Fc-tag
Sequence Human VSIG4, amino acids Met1-Pro283 (Accession # NP_009199.1) with a C-terminal human IgG1 Fc region
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System HEK293 cells
Form Lyophilized
Purity or Quality Grade >95% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.