Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human VSTM2L (NP_542174) VersaClone cDNA
SDP

Catalog No. RDC2111 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDC2111 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. RDC2111 Supplier R&D Systems™ Supplier No. RDC2111
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

R&D Systems™ Human VSTM2L (NP_542174) VersaClone cDNA is a secreted protein that is expressed in the CNS.

The Human VSTM2L (NP_542174) VersaClone cDNA clone contains the complete ORF for VSTM2L, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The VSTM2L insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.

Specifications

Gene VSTM2L
Accession Number NP_542174
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10 mM Tris-Cl
Insert Type 628bp
pH 8.5
Gene ID (Entrez) 128434
Gene Alias C20orf102, chromosome 20 open reading frame 102, dJ1118M15.2, Gm691, V-set and transmembrane domain containing 2 like, V-set and transmembrane domain-containing protein 2-like protein
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Quantity 10 μg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.