Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human WARS Full-length Recombinant Protein with His tag

Catalog No. 89967199 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-967-199 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-967-199 Supplier Abnova™ Supplier No. P5932
Add to Cart
Add to Cart

Used for ELISA, Func, SDS-PAGE, WB

Aminoacyl-tRNA synthetases catalyze the aminoacylation of tRNA by their cognate amino acid. Because of their central role in linking amino acids with nucleotide triplets contained in tRNAs, aminoacyl-tRNA synthetases are thought to be among the first proteins that appeared in evolution. Two forms of tryptophanyl-tRNA synthetase exist, a cytoplasmic form, named WARS, and a mitochondrial form, named WARS2. Tryptophanyl-tRNA synthetase (WARS) catalyzes the aminoacylation of tRNA(trp) with tryptophan and is induced by interferon. Tryptophanyl-tRNA synthetase belongs to the class I tRNA synthetase family. Four transcript variants encoding two different isoforms have been found for this gene. [provided by RefSeq]

Specifications

Concentration 0.67 mg/ml
For Use With (Application) ELISA, Functional Assay, SDS-PAGE, Western Blot
Formulation Liquid
Gene ID (Entrez) 7453
Molecular Weight (g/mol) 53kDa
Name WARS (Human) recombinant protein
Preparation Method Escherichia coli expression system
Purification Method Multi-step chromatography.
Quality Control Testing SDS-PAGE Stained with Coomassie Blue
Quantity 50 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias GAMMA-2/IFI53/IFP53
Common Name WARS
Gene Symbol WARS
Assays Substrate for collagenase
Species E. coli
Recombinant Recombinant
Protein Tag His
Expression System Escherichia coli expression system
Form Liquid
Purity or Quality Grade 0.971
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.