Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human WARS2 Full-length ORF (NP_957715.1, 1 a.a. - 220 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89967206 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-967-206 25 μg
89-967-205 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-967-206 Supplier Abnova™ Supplier No. H00010352P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

Aminoacyl-tRNA synthetases catalyze the aminoacylation of tRNA by their cognate amino acid. Because of their central role in linking amino acids with nucleotide triplets contained in tRNAs, aminoacyl-tRNA synthetases are thought to be among the first proteins that appeared in evolution. Two forms of tryptophanyl-tRNA synthetase exist, a cytoplasmic form, named WARS, and a mitochondrial form, named WARS2. This gene encodes the mitochondrial tryptophanyl-tRNA synthetase. Two alternative transcripts encoding different isoforms have been described. [provided by RefSeq]

Sequence: MALHSMRKARERWSFIRALHKGSAAAPALQKDSKKRVFSGIQPTGILHLGNYLGAIESWVRLQDEYDSVLYSIVDLHSITVPQDPAVLRQSILDMTAVLLACGINPEKSILFQQSQVSEHTQLSWILSCMVRLPRLQHLHQWKAKTTKQKHDGTVGLLTYPVLQAADILLYKSTHVPVGEDQVQHMELVQDLAQGFNKKYGEFFPVPESILSMCVLVFLT

Specifications

Accession Number NP_957715.1
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 10352
Molecular Weight (g/mol) 51.2kDa
Name WARS2 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MALHSMRKARERWSFIRALHKGSAAAPALQKDSKKRVFSGIQPTGILHLGNYLGAIESWVRLQDEYDSVLYSIVDLHSITVPQDPAVLRQSILDMTAVLLACGINPEKSILFQQSQVSEHTQLSWILSCMVRLPRLQHLHQWKAKTTKQKHDGTVGLLTYPVLQAADILLYKSTHVPVGEDQVQHMELVQDLAQGFNKKYGEFFPVPESILSMCVLVFLT
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias TrpRS
Common Name WARS2
Gene Symbol WARS2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.