Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human WASF2 Full-length ORF (NP_008921.1, 1 a.a. - 498 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89967215 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-967-215 10 μg
89-967-216 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-967-215 Supplier Abnova™ Supplier No. H00010163P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a member of the Wiskott-Aldrich syndrome protein family. The gene product is a protein that forms a multiprotein complex that links receptor kinases and actin. Binding to actin occurs through a C-terminal verprolin homology domain in all family members. The multiprotein complex serves to tranduce signals that involve changes in cell shape, motility or function. The published map location (PMID:10381382) has been changed based on recent genomic sequence comparisons, which indicate that the expressed gene is located on chromosome 1, and a pseudogene may be located on chromosome X. [provided by RefSeq]

Sequence: MPLVTRNIEPRHLCRQTLPSVRSELECVTNITLANVIRQLGSLSKYAEDIFGELFTQANTFASRVSSLAERVDRLQVKVTQLDPKEEEVSLQGINTRKAFRSSTIQDQKLFDRNSLPVPVLETYNTCDTPPPLNNLTPYRDDGKEALKFYTDPSYFFDLWKEKMLQDTKDIMKEKRKHRKEKKDNPNRGNVNPRKIKTRKEEWEKMKMGQEFVESKEKLGTSGYPPTLVYQNGSIGCVENVDASSYPPPPQSDSASSPSPSFSEDNLPPPPAEFSYPVDNQRGSGLAGPKRSSVVSPSHPPPAPPLGSPPGPKPGFAPPPAPPPPPPPMIGIPPPPPPVGFGSPGTPPPPSPPSFPPHPDFAAPPPPPPPPAADYPTLPPPPLSQPTGGAPPPPPPPPPPGPPPPPFTGADGQPAIPPPLSDTTKPKSSLPAVSDARSDLLSAIRQGFQLRRVEEQREQEKRDVVGNDVATILSRRIAVEYSDSEDDSSEFDEDDWSD

Specifications

Accession Number NP_008921.1
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 10163
Molecular Weight (g/mol) 80.7kDa
Name WASF2 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MPLVTRNIEPRHLCRQTLPSVRSELECVTNITLANVIRQLGSLSKYAEDIFGELFTQANTFASRVSSLAERVDRLQVKVTQLDPKEEEVSLQGINTRKAFRSSTIQDQKLFDRNSLPVPVLETYNTCDTPPPLNNLTPYRDDGKEALKFYTDPSYFFDLWKEKMLQDTKDIMKEKRKHRKEKKDNPNRGNVNPRKIKTRKEEWEKMKMGQEFVESKEKLGTSGYPPTLVYQNGSIGCVENVDASSYPPPPQSDSASSPSPSFSEDNLPPPPAEFSYPVDNQRGSGLAGPKRSSVVSPSHPPPAPPLGSPPGPKPGFAPPPAPPPPPPPMIGIPPPPPPVGFGSPGTPPPPSPPSFPPHPDFAAPPPPPPPPAADYPTLPPPPLSQPTGGAPPPPPPPPPPGPPPPPFTGADGQPAIPPPLSDTTKPKSSLPAVSDARSDLLSAIRQGFQLRRVEEQREQEKRDVVGNDVATILSRRIAVEYSDSEDDSSEFDEDDWSD
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias SCAR2/WAVE2/dJ393P12.2
Common Name WASF2
Gene Symbol WASF2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.