Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human WDHD1 Partial ORF (NP_009017, 1031 a.a. - 1128 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89967264 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-967-264 10 μg
89-967-265 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-967-264 Supplier Abnova™ Supplier No. H00011169Q01S

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene contains multiple N-terminal WD40 domains and a C-terminal high mobility group (HMG) box. WD40 domains are found in a variety of eukaryotic proteins and may function as adaptor/regulatory modules in signal transduction, pre-mRNA processing and cytoskeleton assembly. HMG boxes are found in many eukaryotic proteins involved in chromatin assembly, transcription and replication. Alternative splicing results in two transcript variants encoding different isoforms. [provided by RefSeq]

Sequence: SNILSDNPDFSDEADIIKEGMIRFRVLSTEERKVWANKAKGETASEGTEAKKRKRVVDESDETENQEEKAKENLNLSKKQKPLDFSTNQKLSAFAFKQ

Specifications

Accession Number NP_009017
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 11169
Molecular Weight (g/mol) 36.52kDa
Name WDHD1 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen SNILSDNPDFSDEADIIKEGMIRFRVLSTEERKVWANKAKGETASEGTEAKKRKRVVDESDETENQEEKAKENLNLSKKQKPLDFSTNQKLSAFAFKQ
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias AND-1
Common Name WDHD1
Gene Symbol WDHD1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.