Learn More
Abnova™ Human WDR23 Full-length ORF (AAH67132.1, 1 a.a. - 546 a.a.) Recombinant Protein with GST-tag at N-terminal
Shop All Abnova Corporation ProductsDescription
This gene encodes a WD repeat-containing protein that interacts with the COP9 signalosome, a macromolecular complex that interacts with cullin-RING E3 ligases and regulates their activity by hydrolyzing cullin-Nedd8 conjugates. Multiple alternatively spliced transcript variants have been found for this gene. [provided by RefSeq]
Specifications
Specifications
| Accession Number | AAH67132.1 |
| For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
| Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
| Gene ID (Entrez) | 80344 |
| Molecular Weight (g/mol) | 88.1kDa |
| Name | WDR23 (Human) Recombinant Protein (P01) |
| Purification Method | Glutathione Sepharose 4 Fast Flow |
| Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
| Quantity | 25 μg |
| Immunogen | MGSRNSSSAGSGSGDPSEGLPRRGAGLRRSEEEEEEDEDVDLAQVLAYLLRRGQVRLVQGGGAANLQFIQALLDSEEENDRAWDGRLGDRYNPPVDATPDTRELEFNEIKTQVELATGQLGLRRAAQKHSFPRMLHQRERGLCHRGSFSLGEQSRVISHFLPNDLGFTDSYSQKAFCGIYSKDGQIFMSACQDQTIRLYDCRYGRFHKFKSIKARDVGWSVLDVAFTPDGNHFLYSSWSDYIHICNIYGEGDTHTALDLRPDERRFAVFSIAVSSDGREVLGGANDGCLYVFDREQNRRTLQIESHEDDVNAVAFADISSQILFSGGDDAICKVWDRRTMREDDPKPVGALAGHQDGITFIDSKGDARYLISNSKDQTIKLWDIRRFSSREGMEASRQAATQQNWDYRWQQVPKKAWRKLKLPGDSSLMTYRGHGVLHTLIRCRFSPIHSTGQQFIYSGCSTGKVVVYDLLSGHIVKKLTNHKACVRDVSWHPFEEKIVSSSWDGNLRLWQYRQAEYFQDDMPESEECASAPAPVPQSSTPFSSPQ |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.