Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human WDR33 Full-length ORF (NP_001006623.1, 1 a.a. - 326 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89967294 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-967-294 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-967-294 Supplier Abnova™ Supplier No. H00055339P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. This gene is highly expressed in testis and the protein is localized to the nucleus. This gene may play important roles in the mechanisms of cytodifferentiation and/or DNA recombination. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene. [provided by RefSeq]

Sequence: MATEIGSPPRFFHMPRFQHQAPRQLFYKRPDFAQQQAMQQLTFDGKRMRKAVNRKTIDYNPSVIKYLENRIWQRDQRDMRAIQPDAGYYNDLVPPIGMLNNPMNAVTTKFVRTSTNKVKCPVFVVRWTPEGRRLVTGASSGEFTLWNGLTFNFETILQAHDSPVRAMTWSHNDMWMLTADHGGYVKYWQSNMNNVKMFQAHKEAIREARFIHNIPFSVVPIVMVKLFSKCILGAEMHGLCQFLGNFLHPINTIFFFVFTHSPFCWHLSEVVLSRYQPLQYVRDVLSAAFCTGFLFSFMINNVYTLFLFIIYCVRQEYFIPNKEFSL

Specifications

Accession Number NP_001006623.1
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 55339
Molecular Weight (g/mol) 64.7kDa
Name WDR33 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MATEIGSPPRFFHMPRFQHQAPRQLFYKRPDFAQQQAMQQLTFDGKRMRKAVNRKTIDYNPSVIKYLENRIWQRDQRDMRAIQPDAGYYNDLVPPIGMLNNPMNAVTTKFVRTSTNKVKCPVFVVRWTPEGRRLVTGASSGEFTLWNGLTFNFETILQAHDSPVRAMTWSHNDMWMLTADHGGYVKYWQSNMNNVKMFQAHKEAIREARFIHNIPFSVVPIVMVKLFSKCILGAEMHGLCQFLGNFLHPINTIFFFVFTHSPFCWHLSEVVLSRYQPLQYVRDVLSAAFCTGFLFSFMINNVYTLFLFIIYCVRQEYFIPNKEFSL
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FLJ11294/WDC146
Common Name WDR33
Gene Symbol WDR33
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.