Learn More
Sino Biological Anti-HE4/WFDC2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 12609RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human WFDC2 / HE4 (rh WFDC2 / HE4; Catalog#12609-H08H; NP_006094.3; Met 1-Phe 124). WFDC2 / HE4 specific IgG was purified by Human WFDC2 / HE4 affinity chromatography.
Catalog No. 50-255-4353
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.