Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human WISP3 Full-length ORF (NP_937882.1, 1 a.a. - 372 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89967418 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-967-418 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-967-418 Supplier Abnova™ Supplier No. H00008838P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a member of the WNT1 inducible signaling pathway (WISP) protein subfamily, which belongs to the connective tissue growth factor (CTGF) family. WNT1 is a member of a family of cysteine-rich, glycosylated signaling proteins that mediate diverse developmental processes. The CTGF family members are characterized by four conserved cysteine-rich domains: insulin-like growth factor-binding domain, von Willebrand factor type C module, thrombospondin domain and C-terminal cystine knot-like domain. This gene is overexpressed in colon tumors. It may be downstream in the WNT1 signaling pathway that is relevant to malignant transformation. Mutations of this gene are associated with progressive pseudorheumatoid dysplasia, an autosomal recessive skeletal disorder, indicating that the gene is essential for normal postnatal skeletal growth and cartilage homeostasis. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MNKRRLLYPSGWLHGPSDMQGLLFSTLLLAGLAQFCCRVQGTGPLDTTPEGRPGEVSDAPQRKQFCHWPCKCPQQKPRCPPGVSLVRDGCGCCKICAKQPGEICNEADLCDPHKGLYCDYSVDRPRYETGVCAYLVAVGCEFNQVHYHNGQVFQPNPLFSCLCVSGAIGCTPLFIPKLAGSHCSGAKGGKKSDQSNCSLEPLLQQLSTSYKTMPAYRNLPLIWKKKCLVQATKWTPCSRTCGMGISNRVTNENSNCEMRKEKRLCYIQPCDSNILKTIKIPKGKTCQPTFQLSKAEKFVFSGCSSTQSYKPTFCGICLDKRCCIPNKSKMITIQFDCPNEGSFKWKMLWITSCVCQRNCREPGDIFSELKIL

Specifications

Accession Number NP_937882.1
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 8838
Molecular Weight (g/mol) 67.8kDa
Name WISP3 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MNKRRLLYPSGWLHGPSDMQGLLFSTLLLAGLAQFCCRVQGTGPLDTTPEGRPGEVSDAPQRKQFCHWPCKCPQQKPRCPPGVSLVRDGCGCCKICAKQPGEICNEADLCDPHKGLYCDYSVDRPRYETGVCAYLVAVGCEFNQVHYHNGQVFQPNPLFSCLCVSGAIGCTPLFIPKLAGSHCSGAKGGKKSDQSNCSLEPLLQQLSTSYKTMPAYRNLPLIWKKKCLVQATKWTPCSRTCGMGISNRVTNENSNCEMRKEKRLCYIQPCDSNILKTIKIPKGKTCQPTFQLSKAEKFVFSGCSSTQSYKPTFCGICLDKRCCIPNKSKMITIQFDCPNEGSFKWKMLWITSCVCQRNCREPGDIFSELKIL
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CCN6/LIBC/MGC125987/MGC125988/MGC125989/PPAC/PPD
Common Name WISP3
Gene Symbol WISP3
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.