Learn More
Gibco™ Human Wnt-7a Recombinant Protein, PeproTech®
Description
120-31-1MG will be provided as 10 x 100 μg (120-31-100UG). 120-31-500UG will be provided as 5 x 100 μg (120-31-100UG). Recombinant Human Wnt-7a is a 35.5 kDa glycoprotein containing 318 amino acids. Due to glycosylation, Wnt-7a migrates between 40-55 kDa by SDS-PAGE gel under unreduced conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Specifications
Specifications
| Accession Number | O00755 |
| For Use With (Application) | Control, Bioactivity |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 7476 |
| Molecular Weight (g/mol) | 35.5 kDa |
| Name | Human Wnt-7a |
| Purification Method | Purified |
| Quantity | 15 μg |
| Source | HEK293 cells |
| Storage Requirements | -20°C |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.