Learn More
Biomatik Corporation Human WNT1 Inducible Signaling Pathway Protein 2 (WISP2) CLIA Kit
Supplier:  Biomatik Corporation EKU10009
Encompass_Preferred
Human WNT1 Inducible Signaling Pathway Protein 2 (WISP2) CLIA Kit for detecting WISP2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 27.4-20,000pg/ml. Sensitivity: 10.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: O76076. Gene ID: 8839. Target Synonyms: CCN5; CT58; CTGF-L; CCN family member 5; Connective tissue growth factor-like protein; Connective tissue growth factor-related protein 58. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5694
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.