Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human WNT10A Full-length ORF (BAG37548.1, 1 a.a. - 417 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89967429 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-967-429 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-967-429 Supplier Abnova™ Supplier No. H00080326P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The WNT gene family consists of structurally related genes which encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. It is strongly expressed in the cell lines of promyelocytic leukemia and Burkitt's lymphoma. In addition, it and another family member, the WNT6 gene, are strongly coexpressed in colorectal cancer cell lines. The gene overexpression may play key roles in carcinogenesis through activation of the WNT-beta-catenin-TCF signaling pathway. This gene and the WNT6 gene are clustered in the chromosome 2q35 region. [provided by RefSeq]

Sequence: MGSAHPRPWLRLRPQPQPRPALWVLLFFLLLLAAAMPRSAPNDILDLRLPPEPVLNANTVCLTLPGLSRRQMEVCVRHPDVAASAIQGIQIAIHECQHQFRDQRWNCSSLETRNKIPYESPIFSRGFRESAFAYAIAAAGVVHAVSNACALGKLKACGCDASRRGDEEAFRRKLHRLQLDALQRGKGLSHGVPEHPALPTASPGLQDSWEWGGCSPDMGFGERFSKDFLDSREPHRDIHARMRLHNNRVGRQAVMENMRRKCKCHGTSGSCQLKTCWQVTPEFRTVGALLRSRFHRATLIRPHNRNGGQLEPGPAGAPSPAPGAPGPRRRASPADLVYFEKSPDFCEREPRLDSAGTVGRLCNKSSAGSDGCGSMCCGRGHNILRQTRSERCHCRFHWCCFVVCEECRITEWVSVCK

Specifications

Accession Number BAG37548.1
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 80326
Molecular Weight (g/mol) 72.27kDa
Name WNT10A (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue
Quantity 2 μg
Immunogen MGSAHPRPWLRLRPQPQPRPALWVLLFFLLLLAAAMPRSAPNDILDLRLPPEPVLNANTVCLTLPGLSRRQMEVCVRHPDVAASAIQGIQIAIHECQHQFRDQRWNCSSLETRNKIPYESPIFSRGFRESAFAYAIAAAGVVHAVSNACALGKLKACGCDASRRGDEEAFRRKLHRLQLDALQRGKGLSHGVPEHPALPTASPGLQDSWEWGGCSPDMGFGERFSKDFLDSREPHRDIHARMRLHNNRVGRQAVMENMRRKCKCHGTSGSCQLKTCWQVTPEFRTVGALLRSRFHRATLIRPHNRNGGQLEPGPAGAPSPAPGAPGPRRRASPADLVYFEKSPDFCEREPRLDSAGTVGRLCNKSSAGSDGCGSMCCGRGHNILRQTRSERCHCRFHWCCFVVCEECRITEWVSVCK
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FLJ14301
Common Name WNT10A
Gene Symbol WNT10A
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.