Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human WNT2B Full-length ORF (NP_078613.1, 1 a.a. - 391 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89967431 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-967-431 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-967-431 Supplier Abnova™ Supplier No. H00007482P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a member of the wingless-type MMTV integration site (WNT) family of highly conserved, secreted signaling factors. WNT family members function in a variety of developmental processes including regulation of cell growth and differentiation and are characterized by a WNT-core domain. This gene may play a role in human development as well as human carcinogenesis. This gene produces two alternatively spliced transcript variants. [provided by RefSeq]

Sequence: MLRPGGAEEAAQLPLRRASAPVPVPSPAAPDGSRASARLGLACLLLLLLLTLPARVDTSWWYIGALGARVICDNIPGLVSRQRQLCQRYPDIMRSVGEGAREWIRECQHQFRHHRWNCTTLDRDHTVFGRVMLRSSREAAFVYAISSAGVVHAITRACSQGELSVCSCDPYTRGRHHDQRGDFDWGGCSDNIHYGVRFAKAFVDAKEKRLKDARALMNLHNNRCGRTAVRRFLKLECKCHGVSGSCTLRTCWRALSDFRRTGDYLRRRYDGAVQVMATQDGANFTAARQGYRRATRTDLVYFDNSPDYCVLDKAAGSLGTAGRVCSKTSKGTDGCEIMCCGRGYDTTRVTRVTQCECKFHWCCAVRCKECRNTVDVHTCKAPKKAEWLDQT

Specifications

Accession Number NP_078613.1
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 7482
Molecular Weight (g/mol) 70.2kDa
Name WNT2B (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MLRPGGAEEAAQLPLRRASAPVPVPSPAAPDGSRASARLGLACLLLLLLLTLPARVDTSWWYIGALGARVICDNIPGLVSRQRQLCQRYPDIMRSVGEGAREWIRECQHQFRHHRWNCTTLDRDHTVFGRVMLRSSREAAFVYAISSAGVVHAITRACSQGELSVCSCDPYTRGRHHDQRGDFDWGGCSDNIHYGVRFAKAFVDAKEKRLKDARALMNLHNNRCGRTAVRRFLKLECKCHGVSGSCTLRTCWRALSDFRRTGDYLRRRYDGAVQVMATQDGANFTAARQGYRRATRTDLVYFDNSPDYCVLDKAAGSLGTAGRVCSKTSKGTDGCEIMCCGRGYDTTRVTRVTQCECKFHWCCAVRCKECRNTVDVHTCKAPKKAEWLDQT
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias WNT13/XWNT2
Common Name WNT2B
Gene Symbol WNT2B
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.