Learn More
Abcam Human Wnt3a Peptide, Synthetic, >90% Purity, 1 mg
Description
- Peptide Type: Synthetic
- Purity: >90%
- Tested Applications: BL
- Form: Lyophilized
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Substrate Properties | Peptide-based Substrate |
| Target Molecule | Wnt3a (Human) |
| Shipping Condition | Blue Ice |
| Form | Lyophilized |
| Label or Dye | Unconjugated |
| Reactivity | Human |
| Content And Storage | Store at -20°C. |
| Cross Reactivity | Human |
| Quantity | 1 mg |
| For Use With (Application) | Blocking, Neutralization |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.