Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human WNT6 Full-length ORF (NP_006513.1, 1 a.a. - 365 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89967444 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-967-444 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-967-444 Supplier Abnova™ Supplier No. H00007475P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The WNT gene family consists of structurally related genes which encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. It is overexpressed in cervical cancer cell line and strongly coexpressed with another family member, WNT10A, in colorectal cancer cell line. The gene overexpression may play key roles in carcinogenesis. This gene and the WNT10A gene are clustered in the chromosome 2q35 region. The protein encoded by this gene is 97% identical to the mouse Wnt6 protein at the amino acid level. [provided by RefSeq]

Sequence: MLPPLPSRLGLLLLLLLCPAHVGGLWWAVGSPLVMDPTSICRKARRLAGRQAELCQAEPEVVAELARGARLGVRECQFQFRFRRWNCSSHSKAFGRILQQDIRETAFVFAITAAGASHAVTQACSMGELLQCGCQAPRGRAPPRPSGLPGTPGPPGPAGSPEGSAAWEWGGCGDDVDFGDEKSRLFMDARHKRGRGDIRALVQLHNNEAGRLAVRSHTRTECKCHGLSGSCALRTCWQKLPPFREVGARLLERFHGASRVMGTNDGKALLPAVRTLKPPGRADLLYAADSPDFCAPNRRTGSPGTRGRACNSSAPDLSGCDLLCCGRGHRQESVQLEENCLCRFHWCCVVQCHRCRVRKELSLCL

Specifications

Accession Number NP_006513.1
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 7475
Molecular Weight (g/mol) 66.1kDa
Name WNT6 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MLPPLPSRLGLLLLLLLCPAHVGGLWWAVGSPLVMDPTSICRKARRLAGRQAELCQAEPEVVAELARGARLGVRECQFQFRFRRWNCSSHSKAFGRILQQDIRETAFVFAITAAGASHAVTQACSMGELLQCGCQAPRGRAPPRPSGLPGTPGPPGPAGSPEGSAAWEWGGCGDDVDFGDEKSRLFMDARHKRGRGDIRALVQLHNNEAGRLAVRSHTRTECKCHGLSGSCALRTCWQKLPPFREVGARLLERFHGASRVMGTNDGKALLPAVRTLKPPGRADLLYAADSPDFCAPNRRTGSPGTRGRACNSSAPDLSGCDLLCCGRGHRQESVQLEENCLCRFHWCCVVQCHRCRVRKELSLCL
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Common Name WNT6
Gene Symbol WNT6
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.