Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human XAGE1 Partial ORF (NP_597674, 19 a.a. - 118 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89967500 Shop All Abnova Corporation Products
Change view
Click to view available options
2μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
89-967-500 2μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-967-500 Supplier Abnova™ Supplier No. H00009503Q01

Used for AP, Array, ELISA, WB-Re

This gene is a member of the XAGE subfamily, which belongs to the GAGE family. The GAGE genes are expressed in a variety of tumors and in some fetal and reproductive tissues. This gene is strongly expressed in Ewing's sarcoma, alveolar rhabdomyosarcoma and normal testis. The protein encoded by this gene contains a nuclear localization signal and shares a sequence similarity with other GAGE/PAGE proteins. Because of the expression pattern and the sequence similarity, this protein also belongs to a family of CT (cancer-testis) antigens. Alternative splicing of this gene, in addition to the use of an alternative transcription start site and an alternative translation initiation codon, results in multiple variants that encode different isoforms. [provided by RefSeq]

Sequence: EGGPRSGGAQAKLGCCWGYPSPRSTWNPDRRFWTPQTGPGEGRHERHTQTQNHTASPRSPVMESPKKKNQQLKVGILHLGSRQKKIRIQLRSQCATWKVI

Specifications

Accession Number NP_597674
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 9503
Molecular Weight (g/mol) 36.74kDa
Name XAGE1 (Human) Recombinant Protein (Q01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 ug
Immunogen EGGPRSGGAQAKLGCCWGYPSPRSTWNPDRRFWTPQTGPGEGRHERHTQTQNHTASPRSPVMESPKKKNQQLKVGILHLGSRQKKIRIQLRSQCATWKVI
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Common Name XAGE1D
Gene Symbol XAGE1D
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.