Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human XBP1 (S Isoform) Alexa Fluor 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC4257T100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC4257T100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC4257T100 Supplier R&D Systems Supplier No. IC4257T100UG
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

XBP1 Monoclonal specifically detects XBP1 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen XBP1
Applications Flow Cytometry
Classification Monoclonal
Clone 525904
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 594
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. NP_001073007
Gene Alias HTF, TREB5, X-box binding protein 1, X-box-binding protein 1, XBP-1, XBP2, XBP2Tax-responsive element-binding protein 5
Gene Symbols XBP1
Host Species Mouse
Immunogen E. coli-derived recombinant human XBP1S Ser188-Leu287 Accession # NP_001073007
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7494
Test Specificity Detects human XBP1 (S Isoform) (XBP1S) in direct ELISAs. This segment is found only in the XBP1S (spliced) variant. Does not detect XBP1U (unspliced).
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.