Learn More
Abcam Human XBP1 Synthetic Peptide, >95% Purity, 50 μg
Description
- Target Protein: XBP1
- Type: Synthetic blocking peptide
- Purity: >95%
- Applications: WB, BL
- Species Reactivity: Human
- Formulation: Liquid, pH 7.2, 0.02% Sodium azide, PBS, 0.1% BSA
- Storage Conditions: Store at -20°C
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Blue Ice |
| Substrate Properties | Peptide-based Substrate |
| Target Molecule | XBP1 (Human) |
| Label or Dye | Unconjugated |
| Form | Liquid |
| Content And Storage | Store at -20°C. |
| Buffer | PBS, 0.1% BSA, 0.02% sodium azide, pH7.2 |
| Concentration | 0.2 mg/mL |
| Cross Reactivity | Human |
| Formulation | PBS, 0.1% BSA, 0.02% Sodium azide, pH 7.2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.