Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human XL Cytokine Luminex™ Performance Panel Premixed Kit

Catalog No. FCSTM18B09
Change view
Click to view available options
Quantity:
10 Plex
11 Plex
12 Plex
13 Plex
14 Plex
15 Plex
16 Plex
17 Plex
18 Plex
19 Plex
2 Plex
20 Plex
21 Plex
22 Plex
23 Plex
24 Plex
25 Plex
26 Plex
27 Plex
28 Plex
29 Plex
3 Plex
30 Plex
31 Plex
32 Plex
33 Plex
34 Plex
35 Plex
36 Plex
37 Plex
38 Plex
39 Plex
4 Plex
40 Plex
41 Plex
42 Plex
43 Plex
44 Plex
45 Plex
5 Plex
6 Plex
7 Plex
8 Plex
9 Plex
44 product options available for selection
Product selection table with 44 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FCSTM18B09 9 Plex
FCSTM18B23 23 Plex
FCSTM18B24 24 Plex
FCSTM18B28 28 Plex
FCSTM18B39 39 Plex
FCSTM18B17 17 Plex
FCSTM18B05 5 Plex
FCSTM18B31 31 Plex
FCSTM18B41 41 Plex
FCSTM18B18 18 Plex
FCSTM18B43 43 Plex
FCSTM18B11 11 Plex
FCSTM18B27 27 Plex
FCSTM18B37 37 Plex
FCSTM18B45 45 Plex
FCSTM18B42 42 Plex
FCSTM18B44 44 Plex
FCSTM18B06 6 Plex
FCSTM18B03 3 Plex
FCSTM18B30 30 Plex
FCSTM18B38 38 Plex
FCSTM18B15 15 Plex
FCSTM18B10 10 Plex
FCSTM18B33 33 Plex
FCSTM18B08 8 Plex
FCSTM18B36 36 Plex
FCSTM18B16 16 Plex
FCSTM18B07 7 Plex
FCSTM18B19 19 Plex
FCSTM18B20 20 Plex
FCSTM18B14 14 Plex
FCSTM18B04 4 Plex
FCSTM18B35 35 Plex
FCSTM18B02 2 Plex
FCSTM18B26 26 Plex
FCSTM18B29 29 Plex
FCSTM18B25 25 Plex
FCSTM18B12 12 Plex
FCSTM18B22 22 Plex
FCSTM18B34 34 Plex
FCSTM18B32 32 Plex
FCSTM18B40 40 Plex
FCSTM18B21 21 Plex
FCSTM18B13 13 Plex
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 44 options available.
44 options
This item is not returnable. View return policy
Catalog No. FCSTM18B09 Supplier R&D Systems™ Supplier No. FCSTM18B09
Add to Cart
Add to Cart
This item is not returnable. View return policy

Specifications

Quantity 9 Plex
Species Human
For Use With (Application) ELISA
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.