Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human XRN1 Full-length ORF (AAH48104.1, 1 a.a. - 459 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89967563 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-967-563 10 μg
89-967-564 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-967-563 Supplier Abnova™ Supplier No. H00054464P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

SEP1 localizes to cytoplasmic foci containing a complex of mRNA-degrading enzymes. In addition to mRNA metabolism, yeast Sep1 has been implicated in a variety of nuclear and cytoplasmic functions, including homologous recombination, meiosis, telomere maintenance, and microtubule assembly.[supplied by OMIM]

Sequence: MGVPKFYRWISERYPCLSEVVKEHQIPEFDNLYLDMNGIIHQCSHPNDDDVHFRISDDKIFTDIFHYLEVLFRIIKPRKVFFMAVDGVAPRAKMNQQRGRRFRSAKEAEDKIKKAIEKGETLPTEARFDSNCITPGTEFMAKLHEHLKYFVNMKISTDKSWQGVTIYFSGHETPGEGEHKIMEFIRSEKAKPDHDPNTRHCLYGLDADLIMLGLTSHEAHFSLLREEVRFGGKKTQRVCAPEETTFHLLHLSLMREYIDYEFSVLKEKITFKYDIERIIDDWILMGFLVGNDFIPHLPHLHINHDALPLLYGTYVTILPELGGYINESGHLNLPRFEKYLVKLSDFDREHFSEVFVDLKWFESKVGNKYLNEAAGVAAEEARNYKEKKKLKGQENSLCWTALDKNEGEMITSKDNLEDETEDDDLFETEFRQYKRTYYMTKMGVDVVSEYVFANAFILK

Specifications

Accession Number AAH48104.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 54464
Molecular Weight (g/mol) 80.2kDa
Name XRN1 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MGVPKFYRWISERYPCLSEVVKEHQIPEFDNLYLDMNGIIHQCSHPNDDDVHFRISDDKIFTDIFHYLEVLFRIIKPRKVFFMAVDGVAPRAKMNQQRGRRFRSAKEAEDKIKKAIEKGETLPTEARFDSNCITPGTEFMAKLHEHLKYFVNMKISTDKSWQGVTIYFSGHETPGEGEHKIMEFIRSEKAKPDHDPNTRHCLYGLDADLIMLGLTSHEAHFSLLREEVRFGGKKTQRVCAPEETTFHLLHLSLMREYIDYEFSVLKEKITFKYDIERIIDDWILMGFLVGNDFIPHLPHLHINHDALPLLYGTYVTILPELGGYINESGHLNLPRFEKYLVKLSDFDREHFSEVFVDLKWFESKVGNKYLNEAAGVAAEEARNYKEKKKLKGQENSLCWTALDKNEGEMITSKDNLEDETEDDDLFETEFRQYKRTYYMTKMGVDVVSEYVFANAFILK
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DKFZp434P0721/DKFZp686B22225/DKFZp686F19113/FLJ41903/SEP1
Common Name XRN1
Gene Symbol XRN1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.