Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human YY1 ELISA Kit

Catalog No. EH488RB
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH488RB 96 Tests
EH488RBX5 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH488RB Supplier Invitrogen™ Supplier No. EH488RB
Add to Cart
Add to Cart

ELISA

Human YY1 quantitates human YY1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human YY1.

YY1 is a ubiquitously distributed transcription factor belonging to the GLI-Kruppel class of zinc finger proteins. The protein is involved in repressing and activating a diverse number of promoters. YY1 may direct histone deacetylases and histone acetyltransferases to a promoter in order to activate or repress the promoter, thus implicating histone modification in the function of YY1.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number AW488674; DELTA; Delta transcription factor; INO80 complex subunit S; INO80S; NF-E1; NMP1; NMP-1; transcriptional repressor protein YY1; Ucrbp; UCRBP transcription factor; UCR-motif DNA-binding protein; yin and yang 1; Yin and Yang 1 protein; Yin Yang 1; YIN-YANG-1; YY1; YY-1; YY1 transcription factor
Assay Range 0.8 to 200 ng/mL
Assay Sensitivity 0.8 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 7528
Gene Symbol YY1
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target YY1
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.