Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ZADH1 Full-length ORF (NP_689657.1, 1 a.a. - 351 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89967654 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-967-654 25 μg
89-967-653 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-967-654 Supplier Abnova™ Supplier No. H00145482P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes an enzyme involved in the metabolism of prostaglandins. The encoded protein catalyzes the NADPH-dependent conversion of 15-keto-prostaglandin E2 to 15-keto-13,14-dihydro-prostaglandin E2. This protein may also be involved in regulating activation of the peroxisome proliferator-activated receptor. Alternative splicing results in multiple transcript variants. [provided by RefSeq]

Sequence: MIVQRVVLNSRPGKNGNPVAENFRMEEVYLPDNINEGQVQVRTLYLSVDPYMRCRMNEDTGTDYITPWQLSQVVDGGGIGIIEESKHTNLTKGDFVTSFYWPWQTKVILDGNSLEKVDPQLVDGHLSYFLGAIGMPGLTSLIGIQEKGHITAGSNKTMVVSGAAGACGSVAGQIGHFLGCSRVVGICGTHEKCILLTSELGFDAAINYKKDNVAEQLRESCPAGVDVYFDNVGGNISDTVISQMNENSHIILCGQISQYNKDVPYPPPLSPAIEAIQKERNITRERFLVLNYKDKFEPGILQLSQWFKEGKLKIKETVINGLENMGAAFQSMMTGGNIGKQIVCISEEISL

Specifications

Accession Number NP_689657.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 145482
Molecular Weight (g/mol) 64.9kDa
Name ZADH1 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MIVQRVVLNSRPGKNGNPVAENFRMEEVYLPDNINEGQVQVRTLYLSVDPYMRCRMNEDTGTDYITPWQLSQVVDGGGIGIIEESKHTNLTKGDFVTSFYWPWQTKVILDGNSLEKVDPQLVDGHLSYFLGAIGMPGLTSLIGIQEKGHITAGSNKTMVVSGAAGACGSVAGQIGHFLGCSRVVGICGTHEKCILLTSELGFDAAINYKKDNVAEQLRESCPAGVDVYFDNVGGNISDTVISQMNENSHIILCGQISQYNKDVPYPPPLSPAIEAIQKERNITRERFLVLNYKDKFEPGILQLSQWFKEGKLKIKETVINGLENMGAAFQSMMTGGNIGKQIVCISEEISL
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DKFZp686P10120/FLJ39091/PGR-2/PGR2/PTRG2/ZADH1
Common Name PTGR2
Gene Symbol PTGR2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.