Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human ZBTB4 Synthetic Peptide

Catalog No. PEP1198
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP1198 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP1198 Supplier Invitrogen™ Supplier No. PEP1198
Add to Cart
Add to Cart

Synthetic Peptide

ZBTB4 Synthetic Peptide, PEP-1198, from Invitrogen.

The ZBTB family of proteins is comprised of diverse zinc finger proteins that also contain a BTB (BR-C, ttk and bab) domain. Similar to Kaiso, a zinc-finger containing protein that can bind methylated CpGs, ZBTB4 can also bind methylated DNA and repress transcription. ZBTB4 has been shown to associate with the Sin3/histone deacetylase co-repressor and repress expression of P21CIP1 as part of a heterodimeric complex with Miz1. In cultured cells, depletion of ZBTB4 promotes cell cycle arrest in response to p53 activation and suppresses apoptosis through regulation of P21CIP1, suggesting that ZBTB4 is a critical determinant of the cellular response to p53 activation. HIPK2, a kinase that is involved in cellular proliferation and survival, phosphorylates and down-regulates ZBTB4 under normal cell growth conditions; this degradation increases with DNA damage.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 57659
Species Human
Purification Method purified
Accession Number Q9P1Z0
Common Name Human ZBTB4
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Symbol ZBTB4
Lyophilized No
Preservative 0.02% sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.