Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ZBTB7B Partial ORF (NP_056956.1, 433 a.a. - 537 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89967709 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-967-709 10 μg
89-967-710 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-967-709 Supplier Abnova™ Supplier No. H00051043Q01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

ZFP67 is an early growth response gene that encodes a zinc finger-containing transcription factor that binds to the promoter regions of type I collagen genes (e.g., COL1A1; MIM 120150) and has a role in development.[supplied by OMIM]

Sequence: HLCHRAFAKEDHLQRHLKGQNCLEVRTRRRRKDDAPPHYPPPSTAAAFPAGLDLSNGHLDTFRLSLARFWEQSAPTWAPVSTPGPPDDDEEEGAPTTPQAEGAME

Specifications

Accession Number NP_056956.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 51043
Molecular Weight (g/mol) 37.29kDa
Name ZBTB7B (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen HLCHRAFAKEDHLQRHLKGQNCLEVRTRRRRKDDAPPHYPPPSTAAAFPAGLDLSNGHLDTFRLSLARFWEQSAPTWAPVSTPGPPDDDEEEGAPTTPQAEGAME
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DKFZp686G01254/THPOK/ZBTB15/ZFP67/ZNF857B/c-Krox/hcKrox
Common Name ZBTB7B
Gene Symbol ZBTB7B
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.