Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human ZC3H12B Synthetic Peptide

Catalog No. PEP1041
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP1041 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP1041 Supplier Invitrogen™ Supplier No. PEP1041
Add to Cart
Add to Cart

Synthetic Peptide

ZC3H12B Synthetic Peptide, PEP-1041, from Invitrogen.

ZC3H12B, also known as MCPIP2, is a member of a family of novel CCCH-zinc finger proteins that regulate macrophage activation and may be involved in host immunity and inflammatory diseases. The function of ZC3H12B is not well understood. Unlike other members of the ZC3H12 family, ZC3H12B has a RasGEF domain at the C-terminal of the zinc finger motif, suggesting its potential involvement in Ras-mediated signal transduction. It is highly expressed in brain, thymus, and testis, but unlike ZC3H12A and ZC3H12C, not expressed in macrophages after treatment with lipopolysaccharide (LPS).
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 340554
Species Human
Purification Method purified
Accession Number Q5HYM0
Common Name Human ZC3H12B
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Symbol ZC3H12B
Lyophilized No
Preservative 0.02% sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.