Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ZC3HAV1 Partial ORF (NP_078901.3, 601 a.a. - 699 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89967734 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-967-734 25 μg
89-967-733 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-967-734 Supplier Abnova™ Supplier No. H00056829Q01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a CCCH-type zinc finger protein that is thought to prevent infection by retroviruses. Studies of the rat homolog indicate that the protein may primarily function to inhibit viral gene expression and induce an innate immunity to viral infection. Alternative splicing occurs at this locus and two variants, each encoding distinct isoforms, are described. [provided by RefSeq]

Sequence: SVFTTKWIWYWKNESGTWIQYGEEKDKRKNSNVDSSYLESLYQSCPRGVVPFQAGSRNYELSFQGMIQTNIASKTQKDVIRRPTFVPQWYVQQMKRGPE

Specifications

Accession Number NP_078901.3
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 56829
Molecular Weight (g/mol) 36.63kDa
Name ZC3HAV1 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen SVFTTKWIWYWKNESGTWIQYGEEKDKRKNSNVDSSYLESLYQSCPRGVVPFQAGSRNYELSFQGMIQTNIASKTQKDVIRRPTFVPQWYVQQMKRGPE
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DKFZp686F2052/DKFZp686H1869/DKFZp686O19171/FLB6421/FLJ13288/MGC48898/ZAP/ZC3H2/ZC3HDC2
Common Name ZC3HAV1
Gene Symbol ZC3HAV1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.