Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ZFHX3 Partial ORF (NP_008816, 2811 a.a. - 2910 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89967826 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-967-826 25 ug
89-967-825 10 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-967-826 Supplier Abnova™ Supplier No. H00000463Q01L

Used for AP, Array, ELISA, WB-Re

This gene encodes a transcription factor with multiple homeodomains and zinc finger motifs, and regulates myogenic and neuronal differentiation. The encoded protein suppresses expression of the alpha-fetoprotein gene by binding to an AT-rich enhancer motif. The protein has also been shown to negatively regulate c-Myb, and transactivate the cell cycle inhibitor cyclin-dependent kinase inhibitor 1A (also known as p21CIP1). This gene is reported to function as a tumor suppressor in several cancers, and sequence variants of this gene are also associated with atrial fibrillation. Multiple transcript variants expressed from alternate promoters and encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: EDFESPSMSSVNLNFDQTKLDNDDCSSVNTAITDTTTGDEGNADNDSATGIATETKSSSAPNEGLTKAAMMAMSEYEDRLSSGLVSPAPSFYSKEYDNEG

Specifications

Accession Number NP_008816
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 463
Molecular Weight (g/mol) 36.74kDa
Name ZFHX3 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 ug
Immunogen EDFESPSMSSVNLNFDQTKLDNDDCSSVNTAITDTTTGDEGNADNDSATGIATETKSSSAPNEGLTKAAMMAMSEYEDRLSSGLVSPAPSFYSKEYDNEG
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias ATBF1/ATBT
Common Name ZFHX3
Gene Symbol ZFHX3
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.