Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ZFYVE9 Full-length ORF (NP_015562.1, 1 a.a. - 762 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89967890 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-967-890 10 μg
89-967-891 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-967-890 Supplier Abnova™ Supplier No. H00009372P02S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a double zinc finger (FYVE domain) protein that interacts directly with SMAD2 and SMAD3, and is involved in Alzheimer's disease. SMAD proteins transmit signals from transmembrane serine/threonine kinase receptors to the nucleus. The FYVE domain has been identified in a number of unrelated signaling molecules. This protein functions to recruit SMAD2 to the transforming growth factor-beta receptor. The FYVE domain is required to maintain the normal localization of this protein but is not involved in mediating interaction with SMADs. The C-terminal domain of this protein interacts with the TGFB receptor. This protein is a component of the TGFB pathway that brings the SMAD substrate to the receptor. Three alternatively spliced transcripts encoding distinct isoforms have been found for this gene. [provided by RefSeq]

Sequence: MENYFQAEAYNLDKVLDEFEQNEDETVSSTLLDTKWNKILDPPSHRLSFNPTLASVNESAVSNESQPQLKVFSLAHSAPLTTEEEDHCANGQDCNLNPEIATMWIDENAVAEDQLIKRNYSWDDQCSAVEVGEKKCGNLACLPDEKNVLVVAVMHNCDKRTLQNDLQDCNNYNSQSLMDAFSCSLDNENRQTDQFSFSINESTEKDMNSEKQMDPLNRPKTEGRSVNHLCPTSSDSLASVCSPSQLKDDGSIGRDPSMSAITSLTVDSVISSQGTDGCPAVKKQENYIPDEDLTGKISSPRTDLGSPNSFSHMSEGILMKKEPAEESTTEESLRSGLPLLLKPDMPNGSGRNNDCERCSDCLVPNEVRADENEGYEHEETLGTTEFLNMTEHFSESQDMTNWKLTKLNEMNDSQVNEEKEKFLQISQPEDTNGDSGGQCVGLADAGLDLKGTCISESEECDFSTVIDTPAANYLSNGCDSYGMQDPGVSFVPKTLPSKEDSVTEEKEIEESKSECYSNIYEQRGNEATEGSGLLLNSTGDLMKKNYLHNFCSQVPSVLGQSSPKVVASLPSISVPFGGARPKQPSNLKLQIPKPLSDHLQNDFPANSGNNTKNKNDILGKAKLGENSATNVCSPSLGNISNVDTNGEHLESYEAEISTRPCLALAPDSPDNDLRAGQFGISARKPFTTLGEVAPVWVPDSQAPNCMKCEARFTFTKRRHHCRACGKVFCASCCSLKCKLLYMDRKEARVCVICHSVLMNGKY

Specifications

Accession Number NP_015562.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 9372
Molecular Weight (g/mol) 110.1kDa
Name ZFYVE9 (Human) Recombinant Protein (P02)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MENYFQAEAYNLDKVLDEFEQNEDETVSSTLLDTKWNKILDPPSHRLSFNPTLASVNESAVSNESQPQLKVFSLAHSAPLTTEEEDHCANGQDCNLNPEIATMWIDENAVAEDQLIKRNYSWDDQCSAVEVGEKKCGNLACLPDEKNVLVVAVMHNCDKRTLQNDLQDCNNYNSQSLMDAFSCSLDNENRQTDQFSFSINESTEKDMNSEKQMDPLNRPKTEGRSVNHLCPTSSDSLASVCSPSQLKDDGSIGRDPSMSAITSLTVDSVISSQGTDGCPAVKKQENYIPDEDLTGKISSPRTDLGSPNSFSHMSEGILMKKEPAEESTTEESLRSGLPLLLKPDMPNGSGRNNDCERCSDCLVPNEVRADENEGYEHEETLGTTEFLNMTEHFSESQDMTNWKLTKLNEMNDSQVNEEKEKFLQISQPEDTNGDSGGQCVGLADAGLDLKGTCISESEECDFSTVIDTPAANYLSNGCDSYGMQDPGVSFVPKTLPSKEDSVTEEKEIEESKSECYSNIYEQRGNEATEGSGLLLNSTGDLMKKNYLHNFCSQVPSVLGQSSPKVVASLPSISVPFGGARPKQPSNLKLQIPKPLSDHLQNDFPANSGNNTKNKNDILGKAKLGENSATNVCSPSLGNISNVDTNGEHLESYEAEISTRPCLALAPDSPDNDLRAGQFGISARKPFTTLGEVAPVWVPDSQAPNCMKCEARFTFTKRRHHCRACGKVFCASCCSLKCKLLYMDRKEARVCVICHSVLMNGKY
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MADHIP/NSP/SARA/SMADIP
Common Name ZFYVE9
Gene Symbol ZFYVE9
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.